Diisopropylamine, 99+%, Thermo Scientific Chemicals
Diisopropylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diisopropylamine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
5 L
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 219432500
Price (USD)/ Each
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-18-9
IUPAC Namebis(propan-2-yl)amine
Molecular FormulaC6H15N
InChI KeyUAOMVDZJSHZZME-UHFFFAOYSA-N
SMILESCC(C)NC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.0 %
Color scale=<10 APHA
Water=<0.2 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only