Thermo Scientific Chemicals

2-Methyl-2,4-pentanediol, 99+%, Thermo Scientific Chemicals

Catalog number: 219450500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Methyl-2,4-pentanediol, 99+%, Thermo Scientific Chemicals

Catalog number: 219450500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 219450500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-41-5
IUPAC Name
2-methylpentane-2,4-diol
Molecular Formula
C6H14O2
InChI Key
SVTBMSDMJJWYQN-UHFFFAOYNA-N
SMILES
CC(O)CC(C)(C)O
Infrared spectrum
Conforms
Refractive index
1.4260 to 1.4280 (20°C, 589 nm)
Color scale
=<15 APHA
Appearance (Form)
Clear liquid
GC
>=99 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text