Acetyl chloride, 99+%, Thermo Scientific Chemicals
Acetyl chloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetyl chloride, 99+%, Thermo Scientific Chemicals

Acetyl chloride, 99+%, C2H3ClO, CAS Number-75-36-5, acetic acid chloride, acetic acid, chloride, acetic chloride, acetylchloride, ccris 4568, ch3cocl, ethanoyl chloride, hsdb 662, rcra waste number u006, unii-qd15rno45k, 250mL, 99% min. (GC), 02, 173, 51.0 deg.C, 100.0, 99.0, CHEBI:37580
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
25 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 219470025
Price (USD)/ Each
293.65
Online exclusive
326.00 
Save 32.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
293.65
Online exclusive
326.00 
Save 32.35 (10%)
Add to cart
Acetyl chloride, 99+%, Thermo Scientific Chemicals
Catalog number219470025
Price (USD)/ Each
293.65
Online exclusive
326.00 
Save 32.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Nameacetyl chloride
Molecular FormulaC2H3ClO
InChI KeyWETWJCDKMRHUPV-UHFFFAOYSA-N
SMILESCC(Cl)=O
Molecular Weight (g/mol)78.50
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
GC>=99.0 %
Titration RCOCl99.0 to 101.0 %
Refractive index1.3875 to 1.3895 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only