Succinic acid, ACS reagent, Thermo Scientific Chemicals
Succinic acid, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Succinic acid, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 219552500
Price (USD)/ Each
45.65
Online exclusive
51.00 
Save 5.35 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
51.00 
Save 5.35 (10%)
Add to cart
Succinic acid, ACS reagent, Thermo Scientific Chemicals
Catalog number219552500
Price (USD)/ Each
45.65
Online exclusive
51.00 
Save 5.35 (10%)
-
Add to cart
Chemical Identifiers
CAS110-15-6
IUPAC Namebutanedioic acid
Molecular FormulaC4H6O4
InChI KeyKDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILESOC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point185.0°C to 191.0°C
Chloride (Cl)=<0.001 %
Sulfate (SO4)=<0.003 %
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only