Thermo Scientific Chemicals

Benzophenone, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 219680010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Benzophenone, 99+%, pure, Thermo Scientific Chemicals

Catalog number: 219680010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 219680010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
119-61-9
IUPAC Name
diphenylmethanone
Molecular Formula
C13H10O
InChI Key
RWCCWEUUXYIKHB-UHFFFAOYSA-N
SMILES
O=C(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White to off-white
GC
>=99.0 %
Appearance (Form)
Crystalline powder or flakes
Infrared spectrum
Conforms
Melting point
47°C to 49°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text