Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzophenone, 99+%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
50 g
250 g
1 kg
Packaging:
View all
Plastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 219680050
Price (USD)/ Each
311.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
366.00 
Save 54.35 (15%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
311.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
366.00 
Save 54.35 (15%)
Add to cart
Benzophenone, 99+%, pure, Thermo Scientific Chemicals
Catalog number219680050
Price (USD)/ Each
311.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
366.00 
Save 54.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namediphenylmethanone
Molecular FormulaC13H10O
InChI KeyRWCCWEUUXYIKHB-UHFFFAOYSA-N
SMILESO=C(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)182.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
GC>=99.0 %
Appearance (Form)Crystalline powder or flakes
Melting point47°C to 49°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only