2,6-Di-tert-butyl-4-methylphenol, 99.8%, Thermo Scientific Chemicals
2,6-Di-tert-butyl-4-methylphenol, 99.8%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Di-tert-butyl-4-methylphenol, 99.8%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 219832500
Price (USD)/ Each
31.65
Online exclusive
35.50 
Save 3.85 (11%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
31.65
Online exclusive
35.50 
Save 3.85 (11%)
Add to cart
2,6-Di-tert-butyl-4-methylphenol, 99.8%, Thermo Scientific Chemicals
Catalog number219832500
Price (USD)/ Each
31.65
Online exclusive
35.50 
Save 3.85 (11%)
-
Add to cart
Chemical Identifiers
CAS128-37-0
IUPAC Name2,6-di-tert-butyl-4-methylphenol
Molecular FormulaC15H24O
InChI KeyNLZUEZXRPGMBCV-UHFFFAOYSA-N
SMILESCC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point69°C to 71°C
Appearance (Color)Semitransparent white
Infrared spectrumConforms
Color of solution=<30 APHA (30 % w/w in acetone)
Water=<0.12 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only