Harshaw TLD™ 90Sr Internal Irradiators
Harshaw TLD&trade; <sup> 90</sup>Sr Internal Irradiators
Thermo Scientific™
Harshaw TLD™ 90Sr Internal Irradiators
Control dosimeter calibration with an internal irradiator for thermoluminescence dosimetry readers.
Have Questions?
Catalog NumberDescription
21989Built-in Internal 90Sr/Y Irradiator
Catalog number 21989
Price (USD)
-
Request A Quote
Description:
Built-in Internal 90Sr/Y Irradiator
Dosimeter calibration is easy to control with the Thermo Scientific™ Harshaw TLD 90Sr Internal Irradiator. The TLD 90Sr Internal Irradiator can be used to calibrate the Thermo Scientific™ Harshaw TLD 6600 and 8800 Automated Reader Instruments through a totally automated process to significantly reduce time and labor costs. The instrument can be calibrated by the user to a NIST™-calibrated source and only one calibration is required for individual element calibration factors, far exceeding batch calibration accuracy.
  • All irradiations are performed under the control of the Reader's firmware; the associated Thermo Scientific™ WinREMS™ Software and PC are not required for this operation.
  • Firmware automatically compensates for normal source decay, eliminating the need for frequent calibrations.
  • Irradiations may be defined by exposure or time.
  • Approximate strength: 0.5mCi nominal exposure rate: 10mR/s.
  • No need for an expensive and large 137Cs source.
  • Dosimeter stability allows extended intervals of several years between recalibration.
Compatible with: 
Thermo Scientific™ Harshaw 6600PLUS Model TLD Reader
Thermo Scientific™ Harshaw 8800PLUS Model TLD Reader
Specifications
DescriptionBuilt-in Internal 90Sr/Y Irradiator
For Use With (Equipment)Thermo Scientific Harshaw 6600 Model TLD Reader; Thermo Scientific Harshaw 8800 Model TLD Reader
Nominal Exposure Rate10 mR/s
Unit SizeEach
Share catalog number, name or link