Thermo Scientific Chemicals

Hydroquinone, 99.5%, Thermo Scientific Chemicals

Catalog number: 219930500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

Hydroquinone, 99.5%, Thermo Scientific Chemicals

Catalog number: 219930500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 219930500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123-31-9
IUPAC Name
benzene-1,4-diol
Molecular Formula
C6H6O2
InChI Key
QIGBRXMKCJKVMJ-UHFFFAOYSA-N
SMILES
OC1=CC=C(O)C=C1
Appearance (Color)
White to off-white
Melting point
171°C to 174°C
Sulfated ash
=<0.05 %
Infrared spectrum
Conforms
Water
=<0.6 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text