Acetic acid, ammonium salt, Thermo Scientific Chemicals
Acetic acid, ammonium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, ammonium salt, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Plastic bottle
Catalog number 219992500
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
Add to cart
Acetic acid, ammonium salt, Thermo Scientific Chemicals
Catalog number219992500
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS631-61-8
IUPAC Nameammonium acetate
Molecular FormulaC2H7NO2
InChI KeyUSFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES[NH4+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
UVat 260 nm A: =<0.04
Appearance (Color)White
pH6.5 to 7.5 (5% soln.in water)
UVat 250 nm A: =<0.05
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only