Thermo Scientific Chemicals

Propionic acid, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 220130010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Propionic acid, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 220130010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 220130010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79-09-4
IUPAC Name
propanoic acid
Molecular Formula
C3H6O2
InChI Key
XBDQKXXYIPTUBI-UHFFFAOYSA-N
SMILES
CCC(O)=O
Appearance (Form)
Clear liquid
Infrared spectrum
Conforms
Refractive index
1.3850 to 1.3870 (20°C, 589 nm)
GC
>=99.0 %
Color scale
=<10 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text