Acetic acid, potassium salt, 99+%, pure, anhydrous, Thermo Scientific Chemicals
Acetic acid, potassium salt, 99+%, pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, potassium salt, 99+%, pure, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
100 g
1 kg
5 kg
Catalog number 220150250
Price (USD)/ Each
920.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1083.00 
Save 162.35 (15%)
-
Add to cart
Quantity:
25 kg
Request bulk or custom format
Price (USD)/ Each
920.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1083.00 
Save 162.35 (15%)
Add to cart
Acetic acid, potassium salt, 99+%, pure, anhydrous, Thermo Scientific Chemicals
Catalog number220150250
Price (USD)/ Each
920.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1083.00 
Save 162.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namepotassium acetate
Molecular FormulaC2H3KO2
InChI KeySCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES[K+].CC([O-])=O
Molecular Weight (g/mol)98.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO4>=99.0 %
Appearance (Form)Crystalline or granular powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only