Thermo Scientific Chemicals

(±)-Propylene oxide, 99.5%, extra pure, Thermo Scientific Chemicals

Catalog number: 220160025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

(±)-Propylene oxide, 99.5%, extra pure, Thermo Scientific Chemicals

Catalog number: 220160025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 220160025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-56-9
IUPAC Name
2-methyloxirane
Molecular Formula
C3H6O
InChI Key
GOOHAUXETOMSMM-UHFFFAOYNA-N
SMILES
CC1CO1
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
Infrared spectrum
Conforms
GC
>=99.4 %
Aldehyde (C3H6O)
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text