Nalgene™ Large Narrow-Mouth Amber Bottles
Nalgene™ Large Narrow-Mouth Amber Bottles
Nalgene™ Large Narrow-Mouth Amber Bottles
Nalgene™ Large Narrow-Mouth Amber Bottles
Nalgene™ Large Narrow-Mouth Amber Bottles
Thermo Scientific™

Nalgene™ Large Narrow-Mouth Amber Bottles

Protect light-sensitive liquids with these amber bottles that reduce UV light transmissions and feature excellent chemical resistance to most acids, bases and alcohols.
Have Questions?
變更視圖buttonViewtableView
產品號碼Capacity (Metric)Closure SizeUnit Size
2204-00052 L38-430 mmCase of 6
2204-00104 L38-430 mmCase of 6
2204-0010PK
亦稱為 2204-0010
4 L38-430 mmEach
2204-00208 L53BCase of 6
2204-0020PK
亦稱為 2204-0020
8 L53BEach
產品號碼 2204-0005
價格 (HKD)/ Case of 6
1,180.00
新增至購物車
Capacity (Metric):
2 L
Closure Size:
38-430 mm
Unit Size:
Case of 6
價格 (HKD)/ Case of 6
1,180.00
新增至購物車

Protect light-sensitive liquids with Thermo Scientific™ Nalgene™ Large Amber Narrow-Mouth Bottles, which reduce UV light transmissions. Useful for storing chemicals and standards, as well as for collecting and storing distilled water, these bottles display excellent chemical resistance to most acids, bases and alcohols.

Features:
  • Opaque amber reduces UV light transmissions to protect light-sensitive liquids
  • Meet the U.S. Pharmacopoeia 23 requirements for light transmission
  • Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles where exposure to light needs to be restricted
  • 4 and 8L sizes feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Autoclavable/Leakproof† when used with Nalgene closures
  • Bottle and carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Note:

Completely disengage threads or remove cap before autoclaving. Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.



Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

規格
Capacity0.5 gal., 2 L
Closure ColorAmber
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38-430 mm
Closure TypePolypropylene Closure
DescriptionAmber Narrow-Mouth Bottle
Diameter (Metric)121 mm
Diameter (Metric) Inner Neck25 mm
Height (English)9.8 in.
Height (Metric)249 mm
Neck Finish38 mm
No. per Case6
No. per Pack1
Through the Closure DispensingNo
Width (English)4.75 in.
AutoclavableAutoclavable
Capacity (English)0.5 gal.
Capacity (Metric)2 L
Certifications/ComplianceComply with U.S. Pharmacopeia XXIII requirements for maximum allowable light transmission
Closure IncludedYes
ColorAmber
GraduatedNo
MaterialPPCO
MouthNarrow
Product TypeNarrow Mouth Bottle
ShapeRound
SterilityNon-sterile
Unit SizeCase of 6

分享目錄編號、產品名稱或連結。