Ethylenediamine, 99+%, Thermo Scientific Chemicals
Ethylenediamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylenediamine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass bottle
Catalog number 220420025
Price (USD)/ Each
118.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
139.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
118.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
139.00 
Save 20.35 (15%)
Add to cart
Ethylenediamine, 99+%, Thermo Scientific Chemicals
Catalog number220420025
Price (USD)/ Each
118.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
139.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
CAS107-15-3
IUPAC Nameethane-1,2-diamine
Molecular FormulaC2H8N2
InChI KeyPIICEJLVQHRZGT-UHFFFAOYSA-N
SMILESNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 % (Coulometric)
Refractive index1.4555 to 1.4575 (20°C, 589 nm)
Color scale=<30 APHA
Infrared spectrumConforms
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only