Thermo Scientific Chemicals

Ethylenediamine, 99+%, Thermo Scientific Chemicals

Catalog number: 220422500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Ethylenediamine, 99+%, Thermo Scientific Chemicals

Catalog number: 220422500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 220422500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
107-15-3
IUPAC Name
ethane-1,2-diamine
Molecular Formula
C2H8N2
InChI Key
PIICEJLVQHRZGT-UHFFFAOYSA-N
SMILES
NCCN
Water
=<0.5 % (Coulometric)
Refractive index
1.4555 to 1.4575 (20°C, 589 nm)
Color scale
=<30 APHA
Infrared spectrum
Conforms
Appearance (Form)
Clear liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text