1x1 image pixel for data collection
Cell MAb Medium, quantum yield
Cell MAb Medium, quantum yield
Gibco™
Cell MAb Medium, quantum yield
Cell MAb Medium, quantum yield, is a chemically defined (CD) basal cell culture medium. It contains L-glutamine and phenol redRead more
Have Questions?
Catalog NumberQuantity
2205111000 mL
Catalog number 220511
Price (USD)
62.25
Each
Add to cart
Quantity:
1000 mL
Price (USD)
62.25
Each
Add to cart
Cell MAb Medium, quantum yield, is a chemically defined (CD) basal cell culture medium. It contains L-glutamine and phenol red and does not contain Pluronic™ F68 (Kolliphor™ P188) or other surfactants. Cell MAb Medium, quantum yield, should be supplemented in the same way as traditional basal medium (e.g., serum or serum-free supplements).
Specifications
Product TypeMAb Medium
Quantity1000 mL
Without AdditivesNo Pluronic Acid, No Other Surfactants
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 8°C. Protect from light.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection