Thermo Scientific Chemicals

Propionaldehyde, 99+%, Thermo Scientific Chemicals

Catalog number: 220515000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Propionaldehyde, 99+%, Thermo Scientific Chemicals

Catalog number: 220515000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 220515000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
123-38-6
IUPAC Name
propanal
Molecular Formula
C3H6O
InChI Key
NBBJYMSMWIIQGU-UHFFFAOYSA-N
SMILES
CCC=O
Refractive index
1.3610 to 1.3660 (20°C, 589 nm)
Infrared spectrum
Conforms
GC
>=99.0 %
Appearance (Form)
Clear liquid
Color scale
=<20 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text