Styrene, 99.5%, for analysis, stabilized
Styrene, 99.5%, for analysis, stabilized
Styrene, 99.5%, for analysis, stabilized
Styrene, 99.5%, for analysis, stabilized
Thermo Scientific Chemicals

Styrene, 99.5%, for analysis, stabilized

CAS: 100-42-5 | C8H8 | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number 220532500
Price (USD)
32.65
Online Exclusive
35.90
Save 3.25 (9%)
Each
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
35.90
Save 3.25 (9%)
Each
Add to cart
Chemical Identifiers
CAS100-42-5
IUPAC Nameethenylbenzene
Molecular FormulaC8H8
InChI KeyPPBRXRYQALVLMV-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5460 to 1.5480 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<20 APHA
Aldehyde=<0.02 % (as benzaldehyde)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection