Thermo Scientific Chemicals

Propylamine, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 220615000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Propylamine, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 220615000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 220615000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-10-8
IUPAC Name
propan-1-amine
Molecular Formula
C3H9N
InChI Key
WGYKZJWCGVVSQN-UHFFFAOYSA-N
SMILES
CCCN
Appearance (Form)
Clear liquid
Water
=<0.2 % (K.F.)
Refractive index
1.3870 to 1.3890 (20°C, 589 nm)
Color scale
=<20 APHA
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text