1-Butanethiol, 99+%, Thermo Scientific Chemicals
1-Butanethiol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Butanethiol, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 220620500
Price (USD)/ Each
24.00
-
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
24.00
Add to cart
1-Butanethiol, 99+%, Thermo Scientific Chemicals
Catalog number220620500
Price (USD)/ Each
24.00
-
Add to cart
Chemical Identifiers
CAS109-79-5
IUPAC Namebutane-1-thiol
Molecular FormulaC4H10S
InChI KeyWQAQPCDUOCURKW-UHFFFAOYSA-N
SMILESCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4420 to 1.4440 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=99.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only