Thermo Scientific Chemicals

Bromoform, 99+%, stabilized, Thermo Scientific Chemicals

Catalog number: 220691000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Bromoform, 99+%, stabilized, Thermo Scientific Chemicals

Catalog number: 220691000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 220691000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
75-25-2
IUPAC Name
tribromomethane
Molecular Formula
CHBr3
InChI Key
DIKBFYAXUHHXCS-UHFFFAOYSA-N
SMILES
BrC(Br)Br
Appearance (Color)
Clear colorless to yellow
Stabilizer
=<1.5 % w/w ethanol
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=99.0 % (excluding stabilizer)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text