Thermo Scientific Chemicals

Dimethyl malonate, 99+%, Thermo Scientific Chemicals

Catalog number: 220700010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Dimethyl malonate, 99+%, Thermo Scientific Chemicals

Catalog number: 220700010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 220700010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
108-59-8
IUPAC Name
1,3-dimethyl propanedioate
Molecular Formula
C5H8O4
InChI Key
BEPAFCGSDWSTEL-UHFFFAOYSA-N
SMILES
COC(=O)CC(=O)OC
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Free acid
=<0.1 % (as malonic acid)
Refractive index
1.4120 to 1.4140 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text