Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 220892500
Price (USD)/ Each
49.65
Online exclusive
54.60 
Save 4.95 (9%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
49.65
Online exclusive
54.60 
Save 4.95 (9%)
Add to cart
Acetic acid, sodium salt, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals
Catalog number220892500
Price (USD)/ Each
49.65
Online exclusive
54.60 
Save 4.95 (9%)
-
Add to cart
Chemical Identifiers
CAS127-09-3
IUPAC Namesodium acetate
Molecular FormulaC2H3NaO2
InChI KeyVMHLLURERBWHNL-UHFFFAOYSA-M
SMILES[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Loss on drying=<1 %
pH7.5 to 9.2 (5 % at 20°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only