Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.
Autoclavable | Not Autoclavable |
Capacity (English) | 4 gal. |
Capacity (Metric) | 15 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White |
Description | Round LDPE Carboy with Handle |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 285 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Height (Metric) With Closure | 429 mm |
Includes | Handles |
Material | LDPE |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Handle |
Shape | Round |
Sterility | Sterile |
Unit Size | Each |
Autoclavable | Not Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White |
Description | Round LDPE Carboy with Handle |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 250 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Height (Metric) With Closure | 389 mm |
Includes | Handles |
Material | LDPE |
No. per Case | 6/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Handle |
Shape | Round |
Sterility | Sterile |
Unit Size | Case of 6 |
Autoclavable | Not Autoclavable |
Capacity (English) | 4 gal. |
Capacity (Metric) | 15 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White |
Description | Round LDPE Carboy with Handle |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 285 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Height (Metric) With Closure | 429 mm |
Includes | Handles |
Material | LDPE |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Handle |
Shape | Round |
Sterility | Sterile |
Unit Size | Case of 4 |
Autoclavable | Not Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White |
Description | Round LDPE Carboy with Handle |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 286 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Height (Metric) With Closure | 528 mm |
Includes | Handles |
Material | LDPE |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Handle |
Shape | Round |
Sterility | Sterile |
Unit Size | Each |
Autoclavable | Not Autoclavable |
Capacity (English) | 6.6 gal. |
Capacity (Metric) | 25 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White |
Description | Round LDPE Carboy with Handle |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 287 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Height (Metric) With Closure | 594 mm |
Includes | Handles |
Material | LDPE |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Handle |
Shape | Round |
Sterility | Sterile |
Unit Size | Case of 4 |
Autoclavable | Not Autoclavable |
Capacity (English) | 13 gal. |
Capacity (Metric) | 50 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White |
Description | Round LDPE Carboy with Handle |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 379 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Height (Metric) With Closure | 678 mm |
Includes | Handles |
Material | LDPE |
Product Type | Carboy with Handle |
Shape | Round |
Sterility | Sterile |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2210-0040PK | Each | 15 L | 151.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2210-0020 | Case of 6 | 10 L | 680.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2210-0040 | Case of 4 | 15 L | 548.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2210-0050PK | Each | 20 L | 173.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2210-0065 | Case of 4 | 25 L | 776.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2210-0130 | Each | 50 L | 349.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Includes:White polypropylene screw cap and 83B closure
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.