Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.
Autoclavable | Not Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | White |
Description | Round LDPE Carboy w/handle |
Diameter (Metric) Inner Neck | 64 mm |
Diameter (Metric) Outer | 286 mm |
For Use With (Application) | Store and transport reagents |
Height (Metric) With Closure | 528 mm |
Includes | Handles |
Material | LDPE |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy with Handle |
Unit Size | Case of 4 |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2210-0050 | Case of 4 | 20 L | 626.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Includes:
White polypropylene screw cap and 83B closure