N-Phenyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals
N-Phenyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Phenyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 221001000
Price (USD)/ Each
37.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
44.50 
Save 6.85 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
37.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
44.50 
Save 6.85 (15%)
Add to cart
N-Phenyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals
Catalog number221001000
Price (USD)/ Each
37.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
44.50 
Save 6.85 (15%)
-
Add to cart
Chemical Identifiers
IUPAC NameN1-phenylbenzene-1,4-diamine
Molecular FormulaC12H12N2
InChI KeyATGUVEKSASEFFO-UHFFFAOYSA-N
SMILESNC1=CC=C(NC2=CC=CC=C2)C=C1
Molecular Weight (g/mol)184.24
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to brown to purple
Appearance (Form)Powder or crystals or flakes or mass and/or chunks
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only