Thermo Scientific Chemicals

N-Phenyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: 221002500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Phenyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: 221002500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 221002500
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
101-54-2
IUPAC Name
N1-phenylbenzene-1,4-diamine
Molecular Formula
C12H12N2
InChI Key
ATGUVEKSASEFFO-UHFFFAOYSA-N
SMILES
NC1=CC=C(NC2=CC=CC=C2)C=C1
Appearance (Color)
Grey to brown to purple
Appearance (Form)
Powder or crystals or flakes or mass and/or chunks
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Skin Sensitization
    Category 1
    Specific target organ toxicity - (single exposure)
    Category 3
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H332 - Harmful if swallowed or if inhaled

    H315 - Causes skin irritation

    H317 - May cause an allergic skin reaction

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear eye protection/ face protection

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P342 + P311 - If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician