Thermo Scientific™

SM(PEG)2 (PEGylated SMCC crosslinker)

Catalog number: 22103
Have Questions?
Thermo Scientific™

SM(PEG)2 (PEGylated SMCC crosslinker)

Catalog number: 22103
Have Questions?
Catalog Number
22103
Unit Size
1 g
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
221031 g
-
22102100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce SM(PEG)n is a series of amine-to-sulfhydryl crosslinkers that differ in length from 17.6 to 95.2 angstroms as a result of polyethylene glycol spacer arms containing n equals 2 to 24 ethylene glycol units.

Features of SM(PEG)2:

Reactive groups: NHS ester and maleimide
Reactive toward: amino and sulfhydryl groups
• Irreversibly crosslink proteins or peptides by flexible PEG spacer arms
• Polyethylene glycol spacer arms help maintain conjugate solubility
• Pure compound with defined structure and molecular weight, ensuring reproducible protein-modification effects
• PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
• Ideal for performing controlled, two-step protein conjugations (see Sulfo-SMCC)

SM(PEG)n NHS- and maleimide-activated PEG compounds for crosslinking between primary amines (NH2) and sulfhydryl (SH) groups in proteins and other molecules. A complete set of compounds are offered, each having the same heterobifunctional structure (NHS-PEGn-Maleimide) but differing in the number of discrete ethylene glycol units ( n = 2, 4, 6, 8, 12 or 24). The N-hydroxysuccinimide ester (NHS) group reacts specifically and efficiently with lysine and N-terminal amino groups at pH 7-9 to form stable amide bonds. The maleimide group reacts with reduced sulfhydryls at pH 6.5-7.5 to form stable thioether bonds.

Alternative Names for SM(PEG)n:
• NHS-dPEG-Mal
• Mal-PEG NHS ester
• NHS-PEO-Maleimide
• Succinimidyl-dPEG-maleimide
• Succinimide-Maleimide PEG

Why use crosslinkers with polyethylene glycol (PEG) spacer arms?
PEG-containing reagents have been used to modify proteins to provide specific advantages. Protein PEGylation can improve the stability of the modified protein, protect it from proteolytic digestion, increase its half life in a biological application, mask it from causing an immunogenic response, decrease its antigenicity or potential toxicity, improve its solubility, diminish the potential for aggregation, and minimize interference for both in vitro and in vivo applications. Polyethylene glycol has these effects because it is nontoxic, nonimmunogenic, hydrophilic, water soluble and highly flexible.

Advantages of discrete-length polyethylene glycol compounds:
These reagents are specially synthesized, resulting in homogeneous compounds of defined molecular weight, characterized by discrete chain lengths, providing a greater ability to optimize and characterize surface protein modifications. Typical preparations of PEG compounds are a heterogeneous mixtures composed of a distribution of chain lengths with a specified average molecular weight or approximate number of PEG subunits.

Related Products
SM(PEG)4 (PEGylated SMCC crosslinker)
SM(PEG)6 (PEGylated, long-chain SMCC crosslinker)
SM(PEG)8 (PEGylated, long-chain SMCC crosslinker)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine-Sulfhydryl
Cleavable
No
Description
SM(PEG)2
Molecular Weight (g/mol)
425.39
PEGylated
Yes
Spacer Arm Length
17.6 Å
Cell Permeability
No
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
Maleimide, NHS Ester
Spacer
Medium (10 to 30 Å)
Form
Solid
Quantity
1 g
Solubility
Water
Format
Standard, Single-use
Water Soluble
Yes
Product Type
Crosslinker

Contents & Storage

Upon receipt store desiccated at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text