Thermo Scientific™

EMCH (N-ε-maleimidocaproic acid hydrazide)

Catalog number: 22106
Have Questions?
Thermo Scientific™

EMCH (N-ε-maleimidocaproic acid hydrazide)

Catalog number: 22106
Have Questions?
Catalog Number
22106
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2210650 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce EMCH is a mid-length, maleimide-and-hydrazide crosslinker for conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as those formed by oxidation of glycoprotein carbohydrates).

Features of EMCH:

Reactive groups: maleimide and hydrazide
Reactive towards: sulfhydryl groups and carbonyl (aldehyde) groups
• Mid-length (11.8Å), sulfhydryl-to-aldehyde crosslinker with simple spacer arm (noncleavable)
• Maleimide group reacts with sulfhydryl groups to form stable thioether linkages
• Hydrazide group conjugates to oxidized sugars of glycoproteins and carbohydrates
• Use sodium meta-periodate to oxidize glycosylation (e.g., sialic acid) to reactive aldehyde groups
• Use with EDC to conjugate primary amine of hydrazide group to carboxyl groups

3,3'-N-[ε-Maleimidocaproic acid] hydrazide, trifluoroacetic acid salt (EMCH) is a water-soluble, heterobifunctional, membrane permeable crosslinker. It contains a sulfhydryl reactive maleimide group and carbonyl reactive hydrazide group at each end of a 6-carbon spacer arm. Maleimides react with sulfhydryls at pH 6.5-7.5 to form stable thiol ether bonds, along with release of the maleimide leaving group. Proteins with cysteine residues not involved in disulfide bond formation are targets for maleimide reactive groups. Carbonyl groups, whilst not found naturally on proteins can be formed by ring sugar reduction to form aldehydes which react with hydrazides at pH5.5-7.5.

Applications:
• Chemical crosslinking of glycoproteins to sulfhydryl-containing molecules
• Use with EDC for reactivity toward carboxyl groups

Specifications of EMCH:
We manufacture EMCH to the highest specifications to produce the most specific bioconjugates, ensure the integrity of your data and to provide you with the highest degree of consistency. Each lot of EMCH is tested to meet the following minimum specifications:

• NMR purity: >90%
• Melting point: 99-110°C
• Identity: IR scan shows only peaks characteristic of the structure and functional groups of EMCH

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
BMPH
MPBH
KMUH
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Carbohydrate-Sulfhydryl
Cleavable
No
Description
EMCH
Molecular Weight (g/mol)
339.29
PEGylated
No
Spacer Arm Length
11.8 Å
Cell Permeability
Yes
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
Amine, Hydrazide, Maleimide
Spacer
Medium (10 to 30 Å)
Form
Solid
Quantity
50 mg
Solubility
Water
Format
Standard
Water Soluble
Yes
Product Type
Crosslinker

Contents & Storage

Upon receipt store desiccated at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text