Tetrabutylammonium fluoride trihydrate, 99%, Thermo Scientific Chemicals
Tetrabutylammonium fluoride trihydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrabutylammonium fluoride trihydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 221082500
Price (USD)/ Each
543.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
639.00 
Save 95.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
543.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
639.00 
Save 95.35 (15%)
Add to cart
Tetrabutylammonium fluoride trihydrate, 99%, Thermo Scientific Chemicals
Catalog number221082500
Price (USD)/ Each
543.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
639.00 
Save 95.35 (15%)
-
Add to cart
Chemical Identifiers
CAS87749-50-6
IUPAC Nametetrabutylazanium trihydrate fluoride
Molecular FormulaC16H42FNO3
InChI KeyVEPTXBCIDSFGBF-UHFFFAOYSA-M
SMILESO.O.O.[F-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only