Thermo Scientific Chemicals

Lead(II,IV) oxide, red, 98%, Thermo Scientific Chemicals

Catalog number: 221110051
5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Lead(II,IV) oxide, red, 98%, Thermo Scientific Chemicals

Catalog number: 221110051
5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 221110051
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1314-41-6
IUPAC Name
bicyclo[3.1.1]triplumboxan-1-yl
Molecular Formula
O4Pb3
InChI Key
XMFOQHDPRMAJNU-UHFFFAOYSA-N
SMILES
O1[Pb]2O[Pb]1O[Pb]O2
Appearance (Form)
Powder
Calcium (Ca)
=<100 ppm
Iron (Fe)
=<50 ppm
Sodium (Na)
=<100 ppm
Potassium (K)
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text