Lead(II,IV) oxide, red, 98%, Thermo Scientific Chemicals
Lead(II,IV) oxide, red, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II,IV) oxide, red, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
5 kg
Packaging:
Glass bottle
Catalog number 221111000
Price (USD)/ Each
25.30
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
25.30
Add to cart
Lead(II,IV) oxide, red, 98%, Thermo Scientific Chemicals
Catalog number221111000
Price (USD)/ Each
25.30
-
Add to cart
Chemical Identifiers
CAS1314-41-6
IUPAC Namebicyclo[3.1.1]triplumboxan-1-yl
Molecular FormulaO4Pb3
InChI KeyXMFOQHDPRMAJNU-UHFFFAOYSA-N
SMILESO1[Pb]2O[Pb]1O[Pb]O2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Calcium (Ca)=<100 ppm
Iron (Fe)=<50 ppm
Sodium (Na)=<100 ppm
Potassium (K)=<100 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only