Thermo Scientific Chemicals

Quinoline, 99%, Thermo Scientific Chemicals

Catalog number: 221140025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Quinoline, 99%, Thermo Scientific Chemicals

Catalog number: 221140025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 221140025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
91-22-5
IUPAC Name
quinoline
Molecular Formula
C9H7N
InChI Key
SMWDFEZZVXVKRB-UHFFFAOYSA-N
SMILES
C1=CC=C2N=CC=CC2=C1
Appearance (Color)
Clear colorless to dark brown
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=98.5 %
Refractive index
1.6230 to 1.6270 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text