Quinoline, 99%, Thermo Scientific Chemicals
Quinoline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Quinoline, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2.5 kg
Packaging:
Glass bottle
Catalog number 221145000
Price (USD)/ Each
158.65
Online exclusive
186.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
158.65
Online exclusive
186.00 
Save 27.35 (15%)
Add to cart
Quinoline, 99%, Thermo Scientific Chemicals
Catalog number221145000
Price (USD)/ Each
158.65
Online exclusive
186.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namequinoline
Molecular FormulaC9H7N
InChI KeySMWDFEZZVXVKRB-UHFFFAOYSA-N
SMILESC1=CC=C2N=CC=CC2=C1
Molecular Weight (g/mol)129.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.6230 to 1.6270 (20°C, 589 nm)
Appearance (Color)Clear colorless to dark brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only