Thermo Scientific Chemicals

3-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 221160050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 221160050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 221160050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18908-07-1
IUPAC Name
1-isocyanato-3-methoxybenzene
Molecular Formula
C8H7NO2
InChI Key
NPOVTGVGOBJZPY-UHFFFAOYSA-N
SMILES
COC1=CC=CC(=C1)N=C=O
Appearance (Form)
Liquid
Refractive index
1.5420 to 1.5440 (20°C, 589 nm)
GC
>=98.5 %
Appearance (Color)
Clear colorless to light pink
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text