1-Bromo-5-chloropentane, 98%, Thermo Scientific Chemicals
1-Bromo-5-chloropentane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-5-chloropentane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 221210250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54512-75-3
IUPAC Name1-bromo-5-chloropentane
Molecular FormulaC5H10BrCl
InChI KeyPHHNNDKXQVKJEP-UHFFFAOYSA-N
SMILESClCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.4826 to 1.4846 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only