Hexacosane, 99%, Thermo Scientific Chemicals
Hexacosane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexacosane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 221220050
Price (USD)/ Each
31.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
36.60 
Save 4.95 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
31.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
36.60 
Save 4.95 (14%)
Add to cart
Hexacosane, 99%, Thermo Scientific Chemicals
Catalog number221220050
Price (USD)/ Each
31.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
36.60 
Save 4.95 (14%)
-
Add to cart
Chemical Identifiers
CAS630-01-3
IUPAC Namehexacosane
Molecular FormulaC26H54
InChI KeyHMSWAIKSFDFLKN-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Crystalline solid
Appearance (Color)White
Melting point56°C to 59°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only