Thermo Scientific Chemicals

Chlorotriisopropylsilane, 97%, Thermo Scientific Chemicals

Catalog number: 221260100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Chlorotriisopropylsilane, 97%, Thermo Scientific Chemicals

Catalog number: 221260100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 221260100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13154-24-0
IUPAC Name
chlorotris(propan-2-yl)silane
Molecular Formula
C9H21ClSi
InChI Key
KQIADDMXRMTWHZ-UHFFFAOYSA-N
SMILES
CC(C)[Si](Cl)(C(C)C)C(C)C
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Refractive index
1.4520 to 1.4540 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text