Chlorotriisopropylsilane, 97%, Thermo Scientific Chemicals
Chlorotriisopropylsilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chlorotriisopropylsilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 221262500
Price (USD)/ Each
753.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
887.00 
Save 133.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
753.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
887.00 
Save 133.35 (15%)
Add to cart
Chlorotriisopropylsilane, 97%, Thermo Scientific Chemicals
Catalog number221262500
Price (USD)/ Each
753.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
887.00 
Save 133.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namechlorotris(propan-2-yl)silane
Molecular FormulaC9H21ClSi
InChI KeyKQIADDMXRMTWHZ-UHFFFAOYSA-N
SMILESCC(C)[Si](Cl)(C(C)C)C(C)C
Molecular Weight (g/mol)192.80
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.4520 to 1.4540 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only