Thermo Scientific Chemicals

Barium perchlorate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 221271000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Barium perchlorate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 221271000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 221271000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13465-95-7
IUPAC Name
barium(2+) diperchlorate
Molecular Formula
BaCl2O8
InChI Key
OOULUYZFLXDWDQ-UHFFFAOYSA-L
SMILES
[Ba++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Titration Complexometric
>=98.5 %
Insoluble matter
=<0.1 % (in water)
Copper (Cu)
=<10 ppm
Nickel (Ni)
=<10 ppm
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text