1x1 image pixel for data collection
Barium perchlorate, 99%, extra pure, anhydrous
Barium perchlorate, 99%, extra pure, anhydrous
Barium perchlorate, 99%, extra pure, anhydrous
Barium perchlorate, 99%, extra pure, anhydrous
Thermo Scientific Chemicals

Barium perchlorate, 99%, extra pure, anhydrous

CAS: 13465-95-7 | BaCl2O8 | 336.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 221275000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13465-95-7
IUPAC Namebarium(2+) diperchlorate
Molecular FormulaBaCl2O8
InChI KeyOOULUYZFLXDWDQ-UHFFFAOYSA-L
SMILES[Ba++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Lead (Pb)=<10 ppm
Chloride (Cl)=<1000 ppm
Iron (Fe)=<10 ppm
Appearance (Form)Powder
Appearance (Color)White
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0461866Certificate of AnalysisApr 11, 2024221275000, 221271000
A0448462Certificate of AnalysisNov 03, 2023221275000, 221271000
A0457328Certificate of AnalysisNov 03, 2023221275000, 221271000
A0403984Certificate of AnalysisDec 13, 2022221275000, 221271000
A0412766Certificate of AnalysisDec 13, 2022221275000, 221271000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection