Copper(II) oxide, 99+%, Thermo Scientific Chemicals
Copper(II) oxide, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) oxide, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 221280025
Price (USD)/ Each
745.65
Online exclusive
877.00 
Save 131.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
745.65
Online exclusive
877.00 
Save 131.35 (15%)
Add to cart
Copper(II) oxide, 99+%, Thermo Scientific Chemicals
Catalog number221280025
Price (USD)/ Each
745.65
Online exclusive
877.00 
Save 131.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namecopper(2+) oxidandiide
Molecular FormulaCuO
InChI KeyKKCXRELNMOYFLS-UHFFFAOYSA-N
SMILES[O--].[Cu++]
Molecular Weight (g/mol)79.55
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=99.0 %
Nitrogen compounds (as N)=<0.002 %
Free alkaliPasses test
Total sulfur (as SO4)=<0.02 %
Chloride (Cl)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only