Thermo Scientific Chemicals

Copper(II) oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 221281000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Copper(II) oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 221281000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 221281000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1317-38-0
IUPAC Name
copper(2+) oxidandiide
Molecular Formula
CuO
InChI Key
KKCXRELNMOYFLS-UHFFFAOYSA-N
SMILES
[O--].[Cu++]
Appearance (Color)
Black
Carbon (C)
=<500 ppm
Appearance (Form)
Powder
Insoluble matter
=<0.02 % (in dilute HCl)
Iron (Fe)
=<500 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text