Thermo Scientific Chemicals

Copper(II) oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 221285000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Copper(II) oxide, 99+%, Thermo Scientific Chemicals

Catalog number: 221285000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 221285000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1317-38-0
IUPAC Name
copper(2+) oxidandiide
Molecular Formula
CuO
InChI Key
KKCXRELNMOYFLS-UHFFFAOYSA-N
SMILES
[O--].[Cu++]
Free alkali
Passes test
Total sulfur (as SO4)
=<0.02 %
Chloride (Cl)
=<50 ppm
Titration Complexometric
>=99.0 %
Nitrogen compounds (as N)
=<0.002 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text