Thermo Scientific Chemicals

Potassium thioacetate, 98%, Thermo Scientific Chemicals

Catalog number: 221305000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium thioacetate, 98%, Thermo Scientific Chemicals

Catalog number: 221305000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 221305000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10387-40-3
IUPAC Name
ethanethioic S-acid potassium
Molecular Formula
C2H4KOS
InChI Key
SDJHDRMYZQFJJO-UHFFFAOYSA-N
SMILES
[K].CC(S)=O
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals and/or chunks
Appearance (Color)
White to light brown
Titration with HClO4
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text