1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole, 98%, Thermo Scientific Chemicals
1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 221310010
Price (USD)/ Each
94.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
111.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
94.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
111.00 
Save 16.35 (15%)
Add to cart
1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole, 98%, Thermo Scientific Chemicals
Catalog number221310010
Price (USD)/ Each
94.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
111.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS74257-00-4
IUPAC Name3-nitro-1-(2,4,6-trimethylbenzenesulfonyl)-1H-1,2,4-triazole
Molecular FormulaC11H12N4O4S
InChI KeySFYDWLYPIXHPML-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Appearance (Color)White to yellow
Infrared spectrumConforms
HPLC>=97.5 %
Melting point131°C to 139°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only