Thermo Scientific Chemicals

2-chloro-5-(chloromethyl)thiophene, 97%, Thermo Scientific Chemicals

Catalog number: 221350010
Each
Thermo Scientific Chemicals

2-chloro-5-(chloromethyl)thiophene, 97%, Thermo Scientific Chemicals

Catalog number: 221350010
Each
Catalog number: 221350010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
23784-96-5
IUPAC Name
2-chloro-5-(chloromethyl)thiophene
Molecular Formula
C5H4Cl2S
InChI Key
MQTKXCOGYOYAMW-UHFFFAOYSA-N
SMILES
ClCC1=CC=C(Cl)S1
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Refractive index
1.5740 to 1.5760 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text