Thermo Scientific Chemicals

Potassium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 221365000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 221365000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 221365000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7778-74-7
IUPAC Name
potassium perchlorate
Molecular Formula
ClKO4
InChI Key
YLMGFJXSLBMXHK-UHFFFAOYSA-M
SMILES
[K+].[O-][Cl](=O)(=O)=O
Appearance (Color)
White
Titration
99.0 to 101.0 %
Chloride (Cl)
=<0.003 %
Iron (Fe)
=<0.0005 %
Sulfate (SO4)
=<0.001 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text