Potassium perchlorate, ACS reagent, Thermo Scientific Chemicals
Potassium perchlorate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium perchlorate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 221375000
Price (USD)/ Each
100.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
118.00 
Save 17.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
100.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
118.00 
Save 17.35 (15%)
Add to cart
Potassium perchlorate, ACS reagent, Thermo Scientific Chemicals
Catalog number221375000
Price (USD)/ Each
100.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
118.00 
Save 17.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7778-74-7
IUPAC Namepotassium perchlorate
Molecular FormulaClKO4
InChI KeyYLMGFJXSLBMXHK-UHFFFAOYSA-M
SMILES[K+].[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Insoluble matter=<0.005 %
Sulfate (SO4)=<0.001 %
Chloride (Cl)=<0.003 %
Calcium (Ca)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only